N-(2-Fluoro-benzyl)-cyclohexane-1,2-diamine hydrochloride

Lieferant: Apollo Scientific

PC408349-500MG
APOSPC408349-500MGEA 430 EUR
APOSPC408349-500MG
N-(2-Fluoro-benzyl)-cyclohexane-1,2-diamine hydrochloride
N-(2-Fluoro-benzyl)-cyclohexane-1,2-diamine hydrochloride
Formel: C₁₃H₁₉FN₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098589
CAS-Nummer: 1353953-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR