(1R,4R)-N1-(5-Fluoro-pyrimidin-2-yl)-cyclohexane-1,4-diamine

Lieferant: Apollo Scientific

PC408350-500MG
APOSPC408350-500MGEA 619 EUR
APOSPC408350-500MG
(1R,4R)-N1-(5-Fluoro-pyrimidin-2-yl)-cyclohexane-1,4-diamine
(1R,4R)-N1-(5-Fluoro-pyrimidin-2-yl)-cyclohexane-1,4-diamine
Formel: C₁₀H₁₅FN₄
Molecular Weight: 210,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098601
CAS-Nummer: 1400637-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR