(1R,4R)-N1-(4-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
Lieferant: Apollo Scientific
PC408351-500MG
APOSPC408351-500MGEA
430
EUR
APOSPC408351-500MG
(1R,4R)-N1-(4-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
(1R,4R)-N1-(4-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
Formel:
C₁₃H₁₉FN₂·ClH Molecular Weight: 258,763 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21098650 CAS-Nummer: 1366386-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...