N,N'-Bis-(4-fluoro-benzyl)-cyclohexane-1,2-diamine
Lieferant: Apollo Scientific
PC408353-500MG
APOSPC408353-500MGEA
430
EUR
APOSPC408353-500MG
N,N'-Bis-(4-fluoro-benzyl)-cyclohexane-1,2-diamine
N,N'-Bis-(4-fluoro-benzyl)-cyclohexane-1,2-diamine
Formel:
C₂₀H₂₄F₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD21098652 CAS-Nummer: 1318762-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...