1-(3-Fluorobenzyl)homopiperazine hydrochloride
Lieferant: Apollo Scientific
Synonyme:
1-(3-Fluorobenzyl)-[1,4]diazepane hydrochloride
PC408354-500MG
APOSPC408354-500MGEA
553
EUR
APOSPC408354-500MG
1-(3-Fluorobenzyl)homopiperazine hydrochloride
1-(3-Fluorobenzyl)homopiperazine hydrochloride
Formel:
C₁₂H₁₇FN₂·ClH Molecular Weight: 244,736 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21098823 CAS-Nummer: 1353965-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...