2-(3-Bromomethyl-piperidin-1-yl)-5-fluoro-pyrimidine
Lieferant: Apollo Scientific
PC408356-500MG
APOSPC408356-500MGEA
619
EUR
APOSPC408356-500MG
2-(3-Bromomethyl-piperidin-1-yl)-5-fluoro-pyrimidine
2-(3-Bromomethyl-piperidin-1-yl)-5-fluoro-pyrimidine
Formel:
C₁₀H₁₃BrFN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD21098891 CAS-Nummer: 1353979-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...