4-Chloromethyl-1-(3-fluoro-benzyl)-piperidine

Lieferant: Apollo Scientific

PC408357-500MG
APOSPC408357-500MGEA 619 EUR
APOSPC408357-500MG
4-Chloromethyl-1-(3-fluoro-benzyl)-piperidine
4-Chloromethyl-1-(3-fluoro-benzyl)-piperidine
Formel: C₁₃H₁₇ClFN
Molecular Weight: 241,732 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18347839
CAS-Nummer: 1341899-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR