3-Bromomethyl-1-(3-fluoro-benzyl)-piperidine

Lieferant: Apollo Scientific

PC408358-500MG
APOSPC408358-500MGEA 619 EUR
APOSPC408358-500MG
3-Bromomethyl-1-(3-fluoro-benzyl)-piperidine
3-Bromomethyl-1-(3-fluoro-benzyl)-piperidine
Formel: C₁₃H₁₇BrFN
Molecular Weight: 286,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098894
CAS-Nummer: 1353948-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR