2-Chloro-1-(3-fluoro-benzyl)-1H-benzoimidazole

Lieferant: Apollo Scientific

PC408359-500MG
APOSPC408359-500MGEA 553 EUR
APOSPC408359-500MG
2-Chloro-1-(3-fluoro-benzyl)-1H-benzoimidazole
2-Chloro-1-(3-fluoro-benzyl)-1H-benzoimidazole
Formel: C₁₄H₁₀ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21098895
CAS-Nummer: 1353979-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR