(S)-1-[1-(3-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-pyrrolidin-3-ol

Lieferant: Apollo Scientific

Synonyme: (S)-1-(1-(3-Fluorobenzyl)-1H-benzo[d]imidazol-2-yl)pyrrolidin-3-ol

PC408360-500MG
APOSPC408360-500MGEA 619 EUR
APOSPC408360-500MG
(S)-1-[1-(3-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-pyrrolidin-3-ol
(S)-1-[1-(3-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-pyrrolidin-3-ol
Formel: C₁₈H₁₈FN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD21098896
CAS-Nummer: 1353998-32-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR