2-(4-Chloro-phenylsulphanyl)-5-fluoro-pyrimidine

Lieferant: Apollo Scientific

Synonyme: 2-((4-Chlorophenyl)thio)-5-fluoropyrimidine

PC408361-500MG
APOSPC408361-500MGEA 0 EUR
APOSPC408361-500MG
2-(4-Chloro-phenylsulphanyl)-5-fluoro-pyrimidine
2-(4-Chloro-phenylsulphanyl)-5-fluoro-pyrimidine
Formel: C₁₀H₆ClFN₂S
Molecular Weight: 240,684 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098907
CAS-Nummer: 1353980-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR