[1-(4-Fluoro-benzenesulphonyl)-piperidin-3-yl]-methyl-amine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 1-((4-Fluorophenyl)sulphonyl)-N-methylpiperidin-3-amine hydrochloride

PC408365-500MG
APOSPC408365-500MGEA 619 EUR
APOSPC408365-500MG
[1-(4-Fluoro-benzenesulphonyl)-piperidin-3-yl]-methyl-amine hydrochloride
[1-(4-Fluoro-benzenesulphonyl)-piperidin-3-yl]-methyl-amine hydrochloride
Formel: C₁₂H₁₇FN₂O₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098933
CAS-Nummer: 1353972-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR