4-Fluoro-N-piperidin-4-ylmethyl-benzenesulphonamide hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-N-(piperidin-4-ylmethyl)benzenesulphonamide hydrochloride

PC408366-500MG
APOSPC408366-500MGEA 619 EUR
APOSPC408366-500MG
4-Fluoro-N-piperidin-4-ylmethyl-benzenesulphonamide hydrochloride
4-Fluoro-N-piperidin-4-ylmethyl-benzenesulphonamide hydrochloride
Formel: C₁₂H₁₇FN₂O₂S·ClH
Molecular Weight: 308,8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098934
CAS-Nummer: 1353966-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR