C-[1-(4-Fluoro-benzenesulphonyl)-piperidin-4-yl]-methylamine hydrochloride

Lieferant: Apollo Scientific

Synonyme: (1-((4-Fluorophenyl)sulphonyl)piperidin-4-yl)methanamine hydrochloride

PC408368-500MG
APOSPC408368-500MGEA 619 EUR
APOSPC408368-500MG
C-[1-(4-Fluoro-benzenesulphonyl)-piperidin-4-yl]-methylamine hydrochloride
C-[1-(4-Fluoro-benzenesulphonyl)-piperidin-4-yl]-methylamine hydrochloride
Formel: C₁₂H₁₇FN₂O₂S·ClH
Molecular Weight: 308,8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098936
CAS-Nummer: 1215609-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR