4-Fluoro-N-piperidin-3-yl-benzenesulphonamide hydrochloride

Lieferant: Apollo Scientific

PC408369-500MG
APOSPC408369-500MGEA 619 EUR
APOSPC408369-500MG
4-Fluoro-N-piperidin-3-yl-benzenesulphonamide hydrochloride
4-Fluoro-N-piperidin-3-yl-benzenesulphonamide hydrochloride
Formel: C₁₁H₁₅FN₂O₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098937
CAS-Nummer: 1353980-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR