C-[1-(4-Fluoro-benzenesulphonyl)-piperidin-3-yl]-methylamine hydrochloride

Lieferant: Apollo Scientific

Synonyme: (1-((4-Fluorophenyl)sulphonyl)piperidin-3-yl)methanamine hydrochloride

PC408370-500MG
APOSPC408370-500MGEA 619 EUR
APOSPC408370-500MG
C-[1-(4-Fluoro-benzenesulphonyl)-piperidin-3-yl]-methylamine hydrochloride
C-[1-(4-Fluoro-benzenesulphonyl)-piperidin-3-yl]-methylamine hydrochloride
Formel: C₁₂H₁₇FN₂O₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098938
CAS-Nummer: 1353985-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR