4-Fluoro-N-piperidin-4-yl-benzenesulphonamide hydrochloride
Lieferant: Apollo Scientific
Synonyme:
4-Fluoro-N-(piperidin-4-yl)benzenesulphonamide hydrochloride
PC408372-500MG
APOSPC408372-500MGEA
619
EUR
APOSPC408372-500MG
4-Fluoro-N-piperidin-4-yl-benzenesulphonamide hydrochloride
4-Fluoro-N-piperidin-4-yl-benzenesulphonamide hydrochloride
Formel:
C₁₁H₁₅FN₂O₂S·ClH Molecular Weight: 294,773 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11099466 CAS-Nummer: 913634-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...