[1-(4-Fluoro-benzenesulphonyl)-piperidin-4-yl]-methyl-amine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 1-((4-Fluorophenyl)sulphonyl)-N-methylpiperidin-4-amine hydrochloride

PC408373-500MG
APOSPC408373-500MGEA 619 EUR
APOSPC408373-500MG
[1-(4-Fluoro-benzenesulphonyl)-piperidin-4-yl]-methyl-amine hydrochloride
[1-(4-Fluoro-benzenesulphonyl)-piperidin-4-yl]-methyl-amine hydrochloride
Formel: C₁₂H₁₇FN₂O₂S·ClH
Molecular Weight: 308,8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098941
CAS-Nummer: 1353958-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR