4-(3-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-(((3-fluorophenyl)thio)methyl)piperidine-1-carboxylate

PC408375-500MG
APOSPC408375-500MGEA 619 EUR
APOSPC408375-500MG
4-(3-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
4-(3-Fluoro-phenylsulphanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₄FNO₂S
Molecular Weight: 325,441 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098955
CAS-Nummer: 1353981-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR