4-(3-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-(((3-Fluorophenyl)thio)methyl)piperidine hydrochloride

PC408376-500MG
APOSPC408376-500MGEA 619 EUR
APOSPC408376-500MG
4-(3-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
4-(3-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
Formel: C₁₂H₁₆FNS·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098956
CAS-Nummer: 1353963-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR