4-Fluoro-N-methyl-N-piperidin-3-yl-benzenesulphonamide hydrochloride

Lieferant: Apollo Scientific

PC408378-500MG
APOSPC408378-500MGEA 619 EUR
APOSPC408378-500MG
4-Fluoro-N-methyl-N-piperidin-3-yl-benzenesulphonamide hydrochloride
4-Fluoro-N-methyl-N-piperidin-3-yl-benzenesulphonamide hydrochloride
Formel: C₁₂H₁₇FN₂O₂S·ClH
Molecular Weight: 308,8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098974
CAS-Nummer: 1353971-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR