4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxymethyl]-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408379-500MG
APOSPC408379-500MGEA 619 EUR
APOSPC408379-500MG
4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxymethyl]-piperidine-1-carboxylic acid tert-butyl ester
4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxymethyl]-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₂₅H₃₀FN₃O₃
Molecular Weight: 439.522 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098981
CAS-Nummer: 1353978-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR