1-(4-Fluoro-benzyl)-2-(piperidin-4-ylmethoxy)-1H-benzoimidazole hydrochloride

Lieferant: Apollo Scientific

PC408380-500MG
APOSPC408380-500MGEA 619 EUR
APOSPC408380-500MG
1-(4-Fluoro-benzyl)-2-(piperidin-4-ylmethoxy)-1H-benzoimidazole hydrochloride
1-(4-Fluoro-benzyl)-2-(piperidin-4-ylmethoxy)-1H-benzoimidazole hydrochloride
Formel: C₂₀H₂₂FN₃O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21098982
CAS-Nummer: 1353966-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR