1-(4-Fluoro-benzyl)-2-(pyrrolidin-3-ylmethoxy)-1H-benzoimidazole hydrochloride

Lieferant: Apollo Scientific

Synonyme: 1-(4-Fluorobenzyl)-2-(pyrrolidin-3-ylmethoxy)-1H-benzo[d]imidazole hydrochloride

PC408381-500MG
APOSPC408381-500MGEA 619 EUR
APOSPC408381-500MG
1-(4-Fluoro-benzyl)-2-(pyrrolidin-3-ylmethoxy)-1H-benzoimidazole hydrochloride
1-(4-Fluoro-benzyl)-2-(pyrrolidin-3-ylmethoxy)-1H-benzoimidazole hydrochloride
Formel: C₁₉H₂₀FN₃O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD21099001
CAS-Nummer: 1353966-61-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR