4-(3-Fluoro-benzenesulphonylmethyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC408383-500MG
APOSPC408383-500MGEA 0 EUR
APOSPC408383-500MG
4-(3-Fluoro-benzenesulphonylmethyl)-piperidine hydrochloride
4-(3-Fluoro-benzenesulphonylmethyl)-piperidine hydrochloride
Formel: C₁₂H₁₆FNO₂S·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22631629
CAS-Nummer: 1417793-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR