1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-3-ol
Lieferant: Apollo Scientific
PC408384-500MG
APOSPC408384-500MGEA
735
EUR
APOSPC408384-500MG
1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-3-ol
1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-3-ol
Formel:
C₁₉H₂₀FN₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD22631647 CAS-Nummer: 1417793-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...