{1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-4-yl}-methanol

Lieferant: Apollo Scientific

Synonyme: (1-(1-(4-Fluorobenzyl)-1H-benzo[d]imidazol-2-yl)piperidin-4-yl)methanol

PC408385-500MG
APOSPC408385-500MGEA 735 EUR
APOSPC408385-500MG
{1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-4-yl}-methanol
{1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-4-yl}-methanol
Formel: C₂₀H₂₂FN₃O
Molecular Weight: 339,407 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631648
CAS-Nummer: 1417794-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR