(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-yl)-methanol

Lieferant: Apollo Scientific

Synonyme: (1-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)piperidin-4-yl)methanol

PC408388-500MG
APOSPC408388-500MGEA 735 EUR
APOSPC408388-500MG
(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-yl)-methanol
(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-4-yl)-methanol
Formel: C₁₂H₁₄ClF₃N₂O
Molecular Weight: 294,701 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13209899
CAS-Nummer: 1308146-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR