(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-yl)-carbamic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408389-500MG
APOSPC408389-500MGEA 735 EUR
APOSPC408389-500MG
(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-yl)-carbamic acid tert-butyl ester
(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-yl)-carbamic acid tert-butyl ester
Formel: C₁₆H₂₁ClF₃N₃O₂
Molecular Weight: 379,805 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631671
CAS-Nummer: 1333866-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR