(S)-1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ol

Lieferant: Apollo Scientific

PC408390-500MG
APOSPC408390-500MGEA 945 EUR
APOSPC408390-500MG
(S)-1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ol
(S)-1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ol
Formel: C₁₀H₁₀ClF₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD22631734
CAS-Nummer: 1311776-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR