3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408391-500MG
APOSPC408391-500MGEA 619 EUR
APOSPC408391-500MG
3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₆H₂₀ClF₃N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22631737
CAS-Nummer: 1417793-56-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR