3-Chloro-2-(pyrrolidin-3-ylmethoxy)-5-trifluoromethyl-pyridine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-2-(pyrrolidin-3-ylmethoxy)-5-(trifluoromethyl)pyridine hydrochloride

PC408392-500MG
APOSPC408392-500MGEA 619 EUR
APOSPC408392-500MG
3-Chloro-2-(pyrrolidin-3-ylmethoxy)-5-trifluoromethyl-pyridine hydrochloride
3-Chloro-2-(pyrrolidin-3-ylmethoxy)-5-trifluoromethyl-pyridine hydrochloride
Formel: C₁₁H₁₂ClF₃N₂O·ClH
Molecular Weight: 317,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631738
CAS-Nummer: 1417794-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR