(S)-3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-pyrrolidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: (S)-tert-Butyl 3-((3-chloro-5-(trifluoromethyl)pyridin-2-yl)oxy)pyrrolidine-1-carboxylate

PC408393-500MG
APOSPC408393-500MGEA 0 EUR
APOSPC408393-500MG
(S)-3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-pyrrolidine-1-carboxylic acid tert-butyl ester
(S)-3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-pyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₅H₁₈ClF₃N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22631739
CAS-Nummer: 1417789-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR