3-Chloro-2-((S)-pyrrolidin-3-yloxy)-5-trifluoromethyl-pyridine hydrochloride

Lieferant: Apollo Scientific

PC408394-500MG
APOSPC408394-500MGEA 865 EUR
APOSPC408394-500MG
3-Chloro-2-((S)-pyrrolidin-3-yloxy)-5-trifluoromethyl-pyridine hydrochloride
3-Chloro-2-((S)-pyrrolidin-3-yloxy)-5-trifluoromethyl-pyridine hydrochloride
Formel: C₁₀H₁₀ClF₃N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22631740
CAS-Nummer: 1417789-61-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR