(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ylmethyl)-carbamic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408395-500MG
APOSPC408395-500MGEA 735 EUR
APOSPC408395-500MG
(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ylmethyl)-carbamic acid tert-butyl ester
(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ylmethyl)-carbamic acid tert-butyl ester
Formel: C₁₇H₂₃ClF₃N₃O₂
Molecular Weight: 393,832 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631676
CAS-Nummer: 1417793-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR