3-Piperidin-4-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine

Lieferant: Apollo Scientific

Synonyme: 3-(Piperidin-4-yl)-2-(trifluoromethyl)-3H-imidazo[4,5-b]pyridine

PC408396-500MG
APOSPC408396-500MGEA 1080 EUR
APOSPC408396-500MG
3-Piperidin-4-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine
3-Piperidin-4-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine
Formel: C₁₂H₁₃F₃N₄
Molecular Weight: 270,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631687
CAS-Nummer: 951544-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR