3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ol

Lieferant: Apollo Scientific

Synonyme: 1-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)piperidin-3-ol

PC408397-500MG
APOSPC408397-500MGEA 735 EUR
APOSPC408397-500MG
3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ol
3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ol
Formel: C₁₁H₁₂ClF₃N₂O
Molecular Weight: 280,674 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13196948
CAS-Nummer: 1292498-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR