1-(3-Fluoro-benzyl)-2-piperazin-1-yl-1H-benzoimidazole

Lieferant: Apollo Scientific

PC408398-500MG
APOSPC408398-500MGEA 865 EUR
APOSPC408398-500MG
1-(3-Fluoro-benzyl)-2-piperazin-1-yl-1H-benzoimidazole
1-(3-Fluoro-benzyl)-2-piperazin-1-yl-1H-benzoimidazole
Formel: C₁₈H₁₉FN₄
Molecular Weight: 310.369 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631777
CAS-Nummer: 1420816-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR