3-[Cyclopropyl-(3-fluoro-benzyl)-carbamoyl]-piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
PC408399-500MG
APOSPC408399-500MGEA
619
EUR
APOSPC408399-500MG
3-[Cyclopropyl-(3-fluoro-benzyl)-carbamoyl]-piperidine-1-carboxylic acid tert-butyl ester
3-[Cyclopropyl-(3-fluoro-benzyl)-carbamoyl]-piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₂₁H₂₉FN₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22631634 CAS-Nummer: 1277958-70-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...