α,2-Dichlor-4-fluortoluol 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Chlor-4-fluorbenzylchlorid

PC4084-1G
APOSPC4084-1GEA 21.9 EUR
APOSPC4084-1G
α,2-Dichlor-4-fluortoluol 96%
α,2-Dichlor-4-fluortoluol
Formel: C₇H₅Cl₂F
Molecular Weight: 179,02 g/mol
Dichte: 1,37 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00236026
CAS-Nummer: 93286-22-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR