4-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-((3-chloro-5-(trifluoromethyl)pyridin-2-yl)oxy)piperidine-1-carboxylate

PC408400-500MG
APOSPC408400-500MGEA 735 EUR
APOSPC408400-500MG
4-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-piperidine-1-carboxylic acid tert-butyl ester
4-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxy)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₆H₂₀ClF₃N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22631664
CAS-Nummer: 1417794-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR