3-Chloro-2-(piperidin-4-yloxy)-5-trifluoromethyl-pyridine hydrochloride

Lieferant: Apollo Scientific

PC408401-500MG
APOSPC408401-500MGEA 0 EUR
APOSPC408401-500MG
3-Chloro-2-(piperidin-4-yloxy)-5-trifluoromethyl-pyridine hydrochloride
3-Chloro-2-(piperidin-4-yloxy)-5-trifluoromethyl-pyridine hydrochloride
Formel: C₁₁H₁₂ClF₃N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22631670
CAS-Nummer: 1417794-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR