[1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-2-yl]-methanol

Lieferant: Apollo Scientific

PC408402-500MG
APOSPC408402-500MGEA 735 EUR
APOSPC408402-500MG
[1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-2-yl]-methanol
[1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-2-yl]-methanol
Formel: C₁₁H₁₂ClF₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD13484412
CAS-Nummer: 1306120-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR