4-[(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-methyl-amino]-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-((3-chloro-5-(trifluoromethyl)pyridin-2-yl)(methyl)amino)piperidine-1-carboxylate

PC408403-500MG
APOSPC408403-500MGEA 735 EUR
APOSPC408403-500MG
4-[(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-methyl-amino]-piperidine-1-carboxylic acid tert-butyl ester
4-[(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-methyl-amino]-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₃ClF₃N₃O₂
Molecular Weight: 393,832 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631680
CAS-Nummer: 1417793-99-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR