3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
PC408404-500MG
APOSPC408404-500MGEA
735
EUR
APOSPC408404-500MG
3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-piperidine-1-carboxylic acid tert-butyl ester
3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₇H₂₂ClF₃N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22631681 CAS-Nummer: 1417793-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...