3-Chloro-2-(piperidin-3-ylmethoxy)-5-trifluoromethyl-pyridine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-2-(piperidin-3-ylmethoxy)-5-(trifluoromethyl)pyridine hydrochloride

PC408405-500MG
APOSPC408405-500MGEA 735 EUR
APOSPC408405-500MG
3-Chloro-2-(piperidin-3-ylmethoxy)-5-trifluoromethyl-pyridine hydrochloride
3-Chloro-2-(piperidin-3-ylmethoxy)-5-trifluoromethyl-pyridine hydrochloride
Formel: C₁₂H₁₄ClF₃N₂O·ClH
Molecular Weight: 331,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631682
CAS-Nummer: 1417793-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR