Ethyl-bis-(3-fluoro-benzyl)-amine

Lieferant: Apollo Scientific

Synonyme: N,N-Bis(3-fluorobenzyl)ethanamine

PC408406-500MG
APOSPC408406-500MGEA 619 EUR
APOSPC408406-500MG
Ethyl-bis-(3-fluoro-benzyl)-amine
Ethyl-bis-(3-fluoro-benzyl)-amine
Formel: C₁₆H₁₇F₂N
Molecular Weight: 261,31 g/mol
MDL: MFCD22631781
CAS-Nummer: 1417794-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR