3-(S)-Pyrrolidin-3-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine

Lieferant: Apollo Scientific

PC408407-500MG
APOSPC408407-500MGEA 1080 EUR
APOSPC408407-500MG
3-(S)-Pyrrolidin-3-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine
3-(S)-Pyrrolidin-3-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine
Formel: C₁₁H₁₁F₃N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22631751
CAS-Nummer: 1417789-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR