1-(3-Fluoro-benzyl)-2-(piperidin-4-ylmethoxy)-1H-benzoimidazole hydrochloride
Lieferant: Apollo Scientific
Synonyme:
1-(3-Fluorobenzyl)-2-(piperidin-4-ylmethoxy)-1H-benzo[d]imidazole hydrochloride
PC408409-500MG
APOSPC408409-500MGEA
735
EUR
APOSPC408409-500MG
1-(3-Fluoro-benzyl)-2-(piperidin-4-ylmethoxy)-1H-benzoimidazole hydrochloride
1-(3-Fluoro-benzyl)-2-(piperidin-4-ylmethoxy)-1H-benzoimidazole hydrochloride
Formel:
C₂₀H₂₂FN₃O·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD22631652 CAS-Nummer: 1417792-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...