Piperidine-3-carboxylic acid cyclopropyl-(3-fluoro-benzyl)-amide hydrochloride

Lieferant: Apollo Scientific

PC408410-500MG
APOSPC408410-500MGEA 735 EUR
APOSPC408410-500MG
Piperidine-3-carboxylic acid cyclopropyl-(3-fluoro-benzyl)-amide hydrochloride
Piperidine-3-carboxylic acid cyclopropyl-(3-fluoro-benzyl)-amide hydrochloride
Formel: C₁₆H₂₁FN₂O·ClH
Molecular Weight: 312,81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631640
CAS-Nummer: 1417793-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR